all cis-5,8,11,14,17-Eicosapentaenoic acid ≥95.0%
Leverantör: TCI EUROPE
Synonyms:
5(Z),8(Z),11(Z),14(Z),17(Z)-Eicosapentaenoic Acid, Timnodonic acid, cis-5,8,11,14,17-Eicosapentaenoic acid, EPA
E0441-100MG
E0441-500MG
TCIAE0441-100MGEA
2225
SEK
TCIAE0441-100MG
TCIAE0441-500MG
all cis-5,8,11,14,17-Eicosapentaenoic acid ≥95.0%
all cis-5,8,11,14,17-Eicosapentaenoic acid
Formel:
C₂₀H₃₀O₂ MW: 302,46 g/mol Flampunkt: 93°C Storage Temperature: Frys |
MDL Number:
MFCD00065716 CAS nummer: 10417-94-4 Merck Index: 14,03531 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...