4,5-Ethylenedithio-1,3-dithiole-2-thione ≥98.0%
Leverantör: TCI EUROPE
E0429-25G
E0429-5G
E0429-1G
TCIAE0429-25GEA
11625
SEK
TCIAE0429-25G
TCIAE0429-5G
TCIAE0429-1G
4,5-Ethylenedithio-1,3-dithiole-2-thione ≥98.0%
4,5-Ethylenedithio-1,3-dithiole-2-thione
Formel:
C₅H₄S₅ MW: 224,39 g/mol |
MDL Number:
MFCD00137882 CAS nummer: 59089-89-3 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 123.0 to 127.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...