Ethyl-2-ethoxybenzoate ≥98.0%
Leverantör: TCI
Synonyms:
2-Ethoxybenzoic Acid Ethyl Ester, Ethyl 2-ethoxybenzoate
E0413-25G
E0413-500G
TCIAE0413-25GEA
1050
SEK
TCIAE0413-25G
TCIAE0413-500G
Ethyl-2-ethoxybenzoate ≥98.0%
Ethyl-2-ethoxybenzoate
Formel:
C₁₁H₁₄O₃ Kokpunkt: 251°C |
MDL Number:
MFCD00142907 CAS nummer: 6290-24-0 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...