N,N,N',N'-Ethylenediaminetetrakis(methylenephosphonic acid) ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: ((Ethane-1,2-diylbis(azanetriyl))tetrakis(methylene))tetraphosphonic acid

E0393-25G E0393-500G E0393-100G
TCIAE0393-25GEA 270 SEK
TCIAE0393-25G TCIAE0393-500G TCIAE0393-100G
N,N,N',N'-Ethylenediaminetetrakis(methylenephosphonic acid) ≥95.0%
N,N,N',N'-Ethylenediaminetetrakis(methylenephosphonic acid)
Formel: C₆H₂₀N₂O₁₂P₄
MW: 436,12 g/mol
MDL Number: MFCD00058893
CAS nummer: 1429-50-1
EINECS: 215-851-5

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 95.0 % (calcd.on anh.substance)
Water max. 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR