N-Ethyl-N-2-hydroxyethyl-m-toluidine ≥96.0%
Leverantör: TCI
Varning
Synonyms: 2-(Ethyl(m-tolyl)amino)ethanol, 2-[Ethyl(3-methylphenyl)amino]ethan-1-ol
E0220-25G E0220-500G
TCIAE0220-25GEA 215 SEK
TCIAE0220-25G TCIAE0220-500G
N-Ethyl-N-2-hydroxyethyl-m-toluidine ≥96.0%
N-Ethyl-N-2-hydroxyethyl-m-toluidine
Formel: C₁₁H₁₇NO
Kokpunkt: 147°C
Flampunkt: 154°C
MDL Number: MFCD00002849
CAS nummer: 91-88-3
EINECS: 202-105-9
Order Now
Specifikation testresultat
Appearance White or Colorless to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 96.0 %