Sodium ethylxanthate ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Sodium Xanthogenate, Ethylxanthic Acid Sodium Salt, Sodium O-Ethyl Dithiocarbonate
E0195-500G
E0195-25G
TCIAE0195-500GEA
7600
SEK
TCIAE0195-500G
TCIAE0195-25G
Sodium ethylxanthate ≥97.0%
Sodium ethylxanthate
Formel:
C₃H₆OS₂ MW: 144,18 g/mol |
MDL Number:
MFCD00068314 CAS nummer: 140-90-9 EINECS: 205-440-9 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (Iodometric Titration) | min. 97.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...