Ethyl vinyl ether ≥98.0% stabilised
Leverantör: TCI EUROPE
Fara
Stabilization: Stabilised with potassium hydroxide
E0193-25ML E0193-500ML E0193-100ML
TCIAE0193-25MLEA 230 SEK
TCIAE0193-25ML TCIAE0193-500ML TCIAE0193-100ML
Ethyl vinyl ether ≥98.0% stabilised
Ethyl vinyl ether
Formel: C₄H₈O
MW: 72,11 g/mol
Kokpunkt: 36°C
Smältpunkt: -115°C
Flampunkt: -45°C
Storage Temperature: Kylskåp
MDL Number: MFCD00009248
CAS nummer: 109-92-2
EINECS: 203-718-4
UN: 1302
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bca42982a3773f1be5530ec8e435e6def8e3978b&vid=bca42982a3773f1be5530ec8e435e6def8e3978b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %