Zinc-N-ethyl-N-phenyldithiocarbamate ≥98.0%

Leverantör: TCI
Varning

E0163-25G E0163-500G
TCIAE0163-25GEA 215 SEK
TCIAE0163-25G TCIAE0163-500G
Zinc-N-ethyl-N-phenyldithiocarbamate ≥98.0%
Zinc-N-ethyl-N-phenyldithiocarbamate
Formel: C₉H₁₁NS₂
MDL Number: MFCD00059686
CAS nummer: 14634-93-6
EINECS: 238-677-1

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Melting point 198.0 to 214.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR