7-Ethyl-2-methyl-4-undecanol ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Tetradecyl Alcohol, Tetradecanol, 7-Ethyl-2-methylundecan-4-ol

E0144-25ML E0144-500ML
TCIAE0144-25MLEA 575 SEK
TCIAE0144-25ML TCIAE0144-500ML
7-Ethyl-2-methyl-4-undecanol ≥98.0%
7-Ethyl-2-methyl-4-undecanol
Formel: C₁₄H₃₀O
MW: 214,39 g/mol
MDL Number: MFCD00027253
CAS nummer: 103-20-8
EINECS: 203-088-0

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR