1,2-Bis(chloroacetoxy)ethane ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Ethyleneglycolbis(monochloroacetatE), Ethylene Glycol Dichloroacetate, Ethylene Glycol Bis(monochloroacetate)

E0107-500G E0107-25G
TCIAE0107-500GEA 4200 SEK
TCIAE0107-500G TCIAE0107-25G
1,2-Bis(chloroacetoxy)ethane ≥98.0%
1,2-Bis(chloroacetoxy)ethane
Formel: C₆H₈Cl₂O₄
MW: 215,03 g/mol
Kokpunkt: 158°C
MDL Number: MFCD00045277
CAS nummer: 6941-69-1
EINECS: 230-096-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82f34b4c8dc71d97cfc538876059fa0b1de14bdb&vid=82f34b4c8dc71d97cfc538876059fa0b1de14bdb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 45.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR