4-Ethoxychrysoidine hydrochloride ≥95.0%

Leverantör: TCI
Varning

E0054-5G E0054-25G E0054-1G
TCIAE0054-5GEA 2200 SEK
TCIAE0054-5G TCIAE0054-25G TCIAE0054-1G
4-Ethoxychrysoidine hydrochloride ≥95.0%
4-Ethoxychrysoidine hydrochloride
Formel: C₁₄H₁₆N₄O
MDL Number: MFCD00042023
CAS nummer: 2313-87-3
EINECS: 219-010-3

Order Now

Specifikation testresultat

Appearance Dark green to Dark red to Black powder to crystal
Purity (HPLC) min. 90.0 area%
Purity (Argentmetric Titration) min. 95.0 %
Transition interval (pH) (red)3.5 to 5.5 (yellow)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR