Natrium 25% (w/w) dispersion i mineralolja, SD Super Fine™
Leverantör: TCI EUROPE
Fara
D5792-100G D5792-25G D5792-5G
TCIAD5792-100GEA 2500 SEK
TCIAD5792-100G TCIAD5792-25G TCIAD5792-5G
Natrium 25% (w/w) dispersion i mineralolja, SD Super Fine™
Natrium
Formel: Na
MW: 22,99 g/mol
Storage Temperature: Frys
MDL Number: MFCD00085307
CAS nummer: 7440-23-5
EINECS: 231-132-9
UN: 1391
ADR: 4.3,I
Merck Index: 14,08570
Order Now
Specifikation testresultat
Appearance Light gray to Brown to Black cloudy liquid
Concentration (Neutralization titration) 25.0 to 27.0 %