DMH-1 ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 4-[6-(4-Isopropoxyphenyl)pyrazolo[1,5-a]pyrimidin-3-yl]quinoline, 4-(6-(4-Isopropoxyphenyl)pyrazolo[1,5-a]pyrimidin-3-yl)quinoline

D5737-10MG D5737-50MG
TCIAD5737-10MGEA 2325 SEK
TCIAD5737-10MG TCIAD5737-50MG
DMH-1 ≥98.0%
DMH-1
Formel: C₂₄H₂₀N₄O
MW: 380,44 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD18384963
CAS nummer: 1206711-16-1

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 176.0 to 180.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR