2,5-Dibromo-3-fluorothiophene ≥98.0%
Leverantör: TCI EUROPE
Varning
D5729-1G D5729-5G
TCIAD5729-1GEA 1900 SEK
TCIAD5729-1G TCIAD5729-5G
2,5-Dibromo-3-fluorothiophene ≥98.0%
2,5-Dibromo-3-fluorothiophene
Formel: C₄HBr₂FS
MW: 259,92 g/mol
Kokpunkt: 78°C
Storage Temperature: Frys
CAS nummer: 32431-85-9
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %