[[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) ≥99.5%, purified by sublimation
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Pyrromethene 546, 5,5-Difluoro-1,3,7,9,10-pentamethyl-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide, BODIPY 493/503
D5724-200MG
D5724-1G
TCIAD5724-200MGEA
765
SEK
TCIAD5724-200MG
TCIAD5724-1G
[[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) ≥99.5%, purified by sublimation
[[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane)
Formel:
C₁₄H₁₇BF₂N₂ MW: 262,11 g/mol Smältpunkt: 257°C |
MDL Number:
MFCD00467372 CAS nummer: 121207-31-6 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 99.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...