trans-2-Dodecenoic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: (E)-Dodec-2-enoic acid
D5660-250MG D5660-1G
TCIAD5660-250MGEA 2000 SEK
TCIAD5660-250MG TCIAD5660-1G
trans-2-Dodecenoic acid ≥98.0% (by GC, titration analysis)
trans-2-Dodecenoic acid
Formel: C₁₂H₂₂O₂
MW: 198,31 g/mol
Kokpunkt: 146°C
Smältpunkt: 23°C
MDL Number: MFCD00045906
CAS nummer: 32466-54-9
Order Now
Specifikation testresultat
Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %