1-(2,5-Xylyl)piperazin ≥98.0% (by GC, titration analysis)
Leverantör: TCI
D5659-25G
D5659-5G
TCIAD5659-25GEA
1250
SEK
TCIAD5659-25G
TCIAD5659-5G
1-(2,5-Xylyl)piperazin ≥98.0% (by GC, titration analysis)
Formel:
C₁₂H₁₈N₂ |
MDL Number:
MFCD00038378 CAS nummer: 1013-25-8 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 44.0 to 48.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...