4'-Demethylepipodophyllotoxin ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: (5R,5aR,8aR,9S)-9-Hydroxy-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one

D5653-1G D5653-5G
TCIAD5653-1GEA 565 SEK
TCIAD5653-1G TCIAD5653-5G
4'-Demethylepipodophyllotoxin ≥98.0%
4'-Demethylepipodophyllotoxin
Formel: C₂₁H₂₀O₈
MW: 400,38 g/mol
Storage Temperature: Frys
MDL Number: MFCD00189421
CAS nummer: 6559-91-7

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -68.0 to -60.0 deg (C=0.65, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR