(Difluoromethyl)bis(2,5-dimethylphenyl)sulphonium tetrafluoroborate ≥95.0% (qNMR)

Leverantör: TCI EUROPE
Fara

D5630-1G D5630-5G
TCIAD5630-1GEA 3025 SEK
TCIAD5630-1G TCIAD5630-5G
(Difluoromethyl)bis(2,5-dimethylphenyl)sulphonium tetrafluoroborate ≥95.0% (qNMR)
(Difluoromethyl)bis(2,5-dimethylphenyl)sulphonium tetrafluoroborate
Formel: C₁₇H₁₉BF₆S
MW: 380,20 g/mol
CAS nummer: 2133476-51-2
UN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f1c367a1b53abb0e90b555984ac7c4be86ca087&vid=a045f9c0ac70d3e435c7b9032a44ecc21056a590&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (1H-qNMR) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR