1',3'-Dihydro-8-methoxy-1',3',3'-trimethyl-6-nitrospiro[2H-1-benzopyran-2,2'-[2H]indole] ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
8-Methoxy-1',3',3'-trimethyl-6-nitrospiro[chromene-2,2'-indole], 8-Methoxy-1',3',3'-trimethyl-6-nitrospiro[chromene-2,2'-indoline], 6-Nitro-8-methoxy BIPS, MNSP
D5626-1G
D5626-5G
TCIAD5626-1GEA
790
SEK
TCIAD5626-1G
TCIAD5626-5G
1',3'-Dihydro-8-methoxy-1',3',3'-trimethyl-6-nitrospiro[2H-1-benzopyran-2,2'-[2H]indole] ≥98.0%
1',3'-Dihydro-8-methoxy-1',3',3'-trimethyl-6-nitrospiro[2H-1-benzopyran-2,2'-[2H]indole]
Formel:
C₂₀H₂₀N₂O₄ MW: 352,39 g/mol |
MDL Number:
MFCD00216608 CAS nummer: 1498-89-1 EINECS: 216-103-0 |
Specifikation testresultat
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...