2,3-Dibromobenzo[b]thiophene ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 2,3-Dibromobenzothiophene
D5604-1G D5604-5G
TCIAD5604-1GEA 495 SEK
TCIAD5604-1G TCIAD5604-5G
2,3-Dibromobenzo[b]thiophene ≥98.0%
2,3-Dibromobenzo[b]thiophene
Formel: C₈H₄Br₂S
MW: 291,9904 g/mol
MDL Number: MFCD06798119
CAS nummer: 6287-82-7
Order Now
Specifikation testresultat
Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C