Dimethyl chlorophosphate ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: Chlorophosphoric acid dimethyl ester, Dimethyl phosphorochloridate

D5601-25G D5601-5G
TCIAD5601-25GEA 2050 SEK
TCIAD5601-25G TCIAD5601-5G
Dimethyl chlorophosphate ≥98.0% (by GC, titration analysis)
Dimethyl chlorophosphate
Formel: C₂H₆ClO₃P
MW: 144,49 g/mol
Kokpunkt: 80°C
Storage Temperature: Frys
MDL Number: MFCD00117904
CAS nummer: 813-77-4
UN: 3265
ADR: 8,II

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR