Desvenlafaxine ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
4-(2-(Dimethylamino)-1-(1-hydroxycyclohexyl)-ethyl)phenol, O-Desmethylvenlafaxine, 4-[2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol, Desvenlafaxine
D5598-1G
D5598-5G
TCIAD5598-1GEA
725
SEK
TCIAD5598-1G
TCIAD5598-5G
Desvenlafaxine ≥98.0%
Desvenlafaxine
Formel:
C₁₆H₂₅NO₂ MW: 263,38 g/mol |
MDL Number:
MFCD00871934 CAS nummer: 93413-62-8 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 220.0 to 224.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...