Dichloro(methyl)(2-phenylethyl)silane ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Dichloro(methyl)(phenethyl)silane

D5569-1G D5569-5G
TCIAD5569-1GEA 405 SEK
TCIAD5569-1G TCIAD5569-5G
Dichloro(methyl)(2-phenylethyl)silane ≥98.0%
Dichloro(methyl)(2-phenylethyl)silane
Formel: C₉H₁₂Cl₂Si
MW: 219,18 g/mol
Kokpunkt: 245°C
Flampunkt: 105°C
MDL Number: MFCD00018084
CAS nummer: 772-65-6
EINECS: 212-253-6
UN: 2987
ADR: 8,II

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR