Methylpropyldichlorosilane ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Silane, dichloromethylpropyl-, dichloro(methyl)(propyl)silane, Dichloro(methyl)propylsilane

D5568-25G D5568-5G
TCIAD5568-25GEA 6750 SEK
TCIAD5568-25G TCIAD5568-5G
Methylpropyldichlorosilane ≥97.0%
Methylpropyldichlorosilane
Formel: C₄H₁₀Cl₂Si
MW: 157,109 g/mol
Kokpunkt: 124°C
Flampunkt: 27°C
MDL Number: MFCD00013608
CAS nummer: 4518-94-9
EINECS: 224-843-0
UN: 2986
ADR: 8,II

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR