Dimethyl Cubane-1,4-dicarboxylate ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
1,4-Bis(methoxycarbonyl)cubane, Cubane-1,4-dicarboxylic acid dimethyl ester
D5542-100MG
D5542-1G
TCIAD5542-100MGEA
1425
SEK
TCIAD5542-100MG
TCIAD5542-1G
Dimethyl Cubane-1,4-dicarboxylate ≥98.0%
Dimethyl cubane-1,4-dicarboxylate
Formel:
C₁₂H₁₂O₄ MW: 220,22 g/mol |
MDL Number:
MFCD29060101 CAS nummer: 29412-62-2 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 163.0 to 167.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...