Mesityleneruthenium(II) chloride Dimer ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Dichloro(mesitylene)ruthenium(II) Dimer
D5524-250MG
D5524-1G
TCIAD5524-250MGEA
1175
SEK
TCIAD5524-250MG
TCIAD5524-1G
Mesityleneruthenium(II) chloride Dimer ≥97.0%
Mesityleneruthenium(II) chloride Dimer
Formel:
C₁₈H₂₄Cl₄Ru₂ MW: 584,33 g/mol |
MDL Number:
MFCD11656079 CAS nummer: 52462-31-4 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...