Diethyl 2,4-Pyridinedicarboxylate ≥97.0%

Leverantör: TCI EUROPE

Synonyms: Diethyl pyridine-2,4-dicarboxylate, 2,4-Pyridinedicarboxylic acid Diethyl ester

D5518-200MG D5518-1G
TCIAD5518-200MGEA 885 SEK
TCIAD5518-200MG TCIAD5518-1G
Diethyl 2,4-Pyridinedicarboxylate ≥97.0%
Diethyl 2,4-Pyridinedicarboxylate
Formel: C₁₁H₁₃NO₄
MW: 223,23 g/mol
Kokpunkt: 180°C
Storage Temperature: Kylskåp
MDL Number: MFCD00152168
CAS nummer: 41438-38-4

Order Now

Specifikation testresultat

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR