2,4-Dichloro-6-phenylpyrimidine ≥98.0%

Leverantör: TCI EUROPE
Varning

D5505-1G D5505-5G
TCIAD5505-1GEA 1600 SEK
TCIAD5505-1G TCIAD5505-5G
2,4-Dichloro-6-phenylpyrimidine ≥98.0%
2,4-Dichloro-6-phenylpyrimidine
Formel: C₁₀H₆Cl₂N₂
MDL Number: MFCD00234591
CAS nummer: 26032-72-4

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 89.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR