1,3,5-Tris[4-[(E)-2-(2,6-di-tert-butylpyrylium-4-yl)vinyl]phenyl]benzene tetrafluoroborate ≥90.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 4,4'-[(1E,1'E)-[5'-[4-[(E)-2-(2,6-Di-tert-butylpyrylium-4-yl)vinyl]phenyl]-[1,1':3',1''-terphenyl]-4,4''-diyl]bis(ethene-2,1-diyl)]bis(2,6-di-tert-butylpyrylium) tetrafluoroborate, 1,3,5-Tris[4-[(E)-2-(2,6-di-tert-butylpyrylium-4-yl)vinyl]phenyl]benzene Tetrafluoroborate

D5497-200MG D5497-1G
TCIAD5497-200MGEA 1550 SEK
TCIAD5497-200MG TCIAD5497-1G
1,3,5-Tris[4-[(E)-2-(2,6-di-tert-butylpyrylium-4-yl)vinyl]phenyl]benzene tetrafluoroborate ≥90.0%
1,3,5-Tris[4-[(E)-2-(2,6-di-tert-butylpyrylium-4-yl)vinyl]phenyl]benzene tetrafluoroborate
Formel: C₆₉H₈₁B₃F₁₂O₃
MW: 1218,81 g/mol
MDL Number: MFCD31618136
CAS nummer: 2056254-18-1
UN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14dd33fe450b2ee8a84ff36c8f16fbf10709af93&vid=d7f388fc4b3cab672ca0632884a51e8252f63a1b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 90.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR