2-Dibenzofuranamine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Aminodibenzofuran, Dibenzo[b,d]furan-2-amine

D5494-1G D5494-5G
TCIAD5494-1GEA 1325 SEK
TCIAD5494-1G TCIAD5494-5G
2-Dibenzofuranamine ≥98.0% (by GC, titration analysis)
2-Dibenzofuranamine
Formel: C₁₂H₉NO
MW: 183,21 g/mol
MDL Number: MFCD00033416
CAS nummer: 3693-22-9

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 127.0 to 131.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR