Diethyl (4-Bromophenyl)phosphonate ≥98.0%

Leverantör: TCI

Synonyms: (4-Bromophenyl)phosphonic acid Diethyl ester

D5453-200MG D5453-1G
TCIAD5453-200MGEA 1300 SEK
TCIAD5453-200MG TCIAD5453-1G
Diethyl (4-Bromophenyl)phosphonate ≥98.0%
Diethyl (4-Bromophenyl)phosphonate
Formel: C₁₀H₁₄BrO₃P
Kokpunkt: 120°C
MDL Number: MFCD00769296
CAS nummer: 20677-12-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=370979bde2a9c80ab65c52457f661722543e98d2&vid=370979bde2a9c80ab65c52457f661722543e98d2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR