Dacomitinib ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
(E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-methoxyquinazolin-6-yl]-4-(piperidin-1-yl)but-2-enamide, (E)-N-(4-((3-Chloro-4-fluorophenyl)amino)-7-methoxyquinazolin-6-yl)-4-(piperidin-1-yl)but-2-enamide, PF 00299804
D5450-250MG
D5450-50MG
TCIAD5450-250MGEA
6175
SEK
TCIAD5450-250MG
TCIAD5450-50MG
Dacomitinib ≥98.0%
Dacomitinib
Formel:
C₂₄H₂₅ClFN₅O₂ MW: 469,95 g/mol Storage Temperature: Frys |
MDL Number:
MFCD19443734 CAS nummer: 1110813-31-4 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 14.00 to 16.00 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...