(2,3-Dihydrothieno[3,4-b][1,4]dioxin-2-yl)methanol ≥97.0%
Leverantör: TCI
Synonyms: Hydroxymethyl EDOT, EDT-methanol
D5447-1G D5447-200MG
TCIAD5447-1GEA 2850 SEK
TCIAD5447-1G TCIAD5447-200MG
(2,3-Dihydrothieno[3,4-b][1,4]dioxin-2-yl)methanol ≥97.0%
(2,3-Dihydrothieno[3,4-b][1,4]dioxin-2-yl)methanol
Formel: C₇H₈O₃S
Kokpunkt: 120°C
Storage Temperature: Kylskåp
MDL Number: MFCD11045357
CAS nummer: 146796-02-3
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %