5,5'-Diiodo-2,2'-dimethylbiphenyl ≥97.0%
Leverantör: TCI EUROPE
D5430-1G
D5430-5G
TCIAD5430-1GEA
1600
SEK
TCIAD5430-1G
TCIAD5430-5G
5,5'-Diiodo-2,2'-dimethylbiphenyl ≥97.0%
5,5'-Diiodo-2,2'-dimethylbiphenyl
Formel:
C₁₄H₁₂I₂ MW: 434,06 g/mol |
CAS nummer:
1392203-19-8 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 115.0 to 119.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...