2,6-Dichloroquinoline ≥98.0%

Leverantör: TCI
Varning

D5429-1G D5429-5G
TCIAD5429-1GEA 565 SEK
TCIAD5429-1G TCIAD5429-5G
2,6-Dichloroquinoline ≥98.0%
2,6-Dichloroquinoline
Formel: C₉H₅Cl₂N
MW: 198.0500 g/mol
Kokpunkt: 172°C
MDL Number: MFCD03427194
CAS nummer: 1810-72-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48449af29deafb8e411ddbf8c602b5e5f155b192&vid=48449af29deafb8e411ddbf8c602b5e5f155b192&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 160.0 to 164.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR