2,7-Diphenyl-9H-carbazole ≥98.0%
Leverantör: TCI
|
Varning
|
D5427-1G
D5427-5G
TCIAD5427-1GEA
1100
SEK
TCIAD5427-1G
TCIAD5427-5G
2,7-Diphenyl-9H-carbazole ≥98.0%
2,7-Diphenyl-9H-carbazole
Formel:
C₂₄H₁₇N |
MDL Number:
MFCD30478534 CAS nummer: 42448-04-4 |
|
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...