1,3-Di(2-pyridyl)propane ≥97.0%
Leverantör: TCI EUROPE
D5415-25G D5415-500G
TCIAD5415-25GEA 1050 SEK
TCIAD5415-25G TCIAD5415-500G
1,3-Di(2-pyridyl)propane ≥97.0%
1,3-Di(2-pyridyl)propane
Formel: C₁₃H₁₄N₂
MW: 198,27 g/mol
Kokpunkt: 117°C
CAS nummer: 15937-81-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e2e91f1207bf0322a842fb87c55ed49257ca690&vid=6e2e91f1207bf0322a842fb87c55ed49257ca690&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white clear liquid
Purity (GC) min. 97.0 %