3,6-Dibromo-9-hexyl-9H-carbazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
D5410-1G
D5410-5G
TCIAD5410-1GEA
1275
SEK
TCIAD5410-1G
TCIAD5410-5G
3,6-Dibromo-9-hexyl-9H-carbazole ≥98.0%
3,6-Dibromo-9-hexyl-9H-carbazole
Formel:
C₁₈H₁₉Br₂N MW: 409,17 g/mol |
MDL Number:
MFCD23099108 CAS nummer: 150623-72-6 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...