2,7-Dibromo-9-heptyl-9H-carbazole ≥98.0%

Leverantör: TCI
Varning

D5403-1G D5403-5G
TCIAD5403-1GEA 1325 SEK
TCIAD5403-1G TCIAD5403-5G
2,7-Dibromo-9-heptyl-9H-carbazole ≥98.0%
2,7-Dibromo-9-heptyl-9H-carbazole
Formel: C₁₉H₂₁Br₂N
MDL Number: MFCD30742903
CAS nummer: 1173071-58-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50a74c0f9f3cdeec49716f88a49534b284ad6cef&vid=50a74c0f9f3cdeec49716f88a49534b284ad6cef&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR