3,6-Dibromo-9-(4-dodecyloxyphenyl)-9H-carbazole ≥97.0% (by HPLC, total nitrogen)

Leverantör: TCI
Varning

D5343-200MG D5343-1G
TCIAD5343-200MGEA 855 SEK
TCIAD5343-200MG TCIAD5343-1G
3,6-Dibromo-9-(4-dodecyloxyphenyl)-9H-carbazole ≥97.0% (by HPLC, total nitrogen)
3,6-Dibromo-9-(4-dodecyloxyphenyl)-9H-carbazole
Formel: C₃₀H₃₅Br₂NO
MW: 585.42 g/mol
MDL Number: MFCD30536722
CAS nummer: 865163-47-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2799e233af87add3e38be9c8a31eb1b425f3320&vid=c2799e233af87add3e38be9c8a31eb1b425f3320&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 89.0 to 93.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR