3,6-Dibromo-9-[4-(2-ethylhexyloxy)phenyl]-9H-carbazole ≥95.0% (by HPLC, total nitrogen)
Leverantör: TCI
|
Varning
|
D5342-200MG
D5342-1G
TCIAD5342-200MGEA
725
SEK
TCIAD5342-200MG
TCIAD5342-1G
3,6-Dibromo-9-[4-(2-ethylhexyloxy)phenyl]-9H-carbazole ≥95.0% (by HPLC, total nitrogen)
3,6-Dibromo-9-[4-(2-ethylhexyloxy)phenyl]-9H-carbazole
Formel:
C₂₆H₂₇Br₂NO MW: 529.32 g/mol |
MDL Number:
MFCD30721573 CAS nummer: 946491-48-1 |
|
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (with Total Nitrogen) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...