3,6-Dibromo-9-(4-n-octyloxyphenyl)-9H-carbazole ≥98.0%

Leverantör: TCI
Varning

Synonyms: 3,6-Dibromo-9-(4-octoxyphenyl)carbazole

D5341-200MG D5341-1G
TCIAD5341-200MGEA 845 SEK
TCIAD5341-200MG TCIAD5341-1G
3,6-Dibromo-9-(4-n-octyloxyphenyl)-9H-carbazole ≥98.0%
3,6-Dibromo-9-(4-n-octyloxyphenyl)-9H-carbazole
Formel: C₂₆H₂₇Br₂NO
MW: 529.32 g/mol
MDL Number: MFCD30536723
CAS nummer: 917773-26-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e28661aadb5ef070fce83e6633842f7061478d2&vid=4e28661aadb5ef070fce83e6633842f7061478d2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 76.0 to 80.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR