(3,5-Dibromophenyl)triphenylsilane ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 3,5-Dibromotetraphenylsilane

D5338-200MG D5338-1G
TCIAD5338-200MGEA 925 SEK
TCIAD5338-200MG TCIAD5338-1G
(3,5-Dibromophenyl)triphenylsilane ≥98.0%
(3,5-Dibromophenyl)triphenylsilane
Formel: C₂₄H₁₈Br₂Si
MW: 494,30 g/mol
CAS nummer: 1030856-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bb19099fd6a3c93e59d9526d712c6cf38e02708&vid=8bb19099fd6a3c93e59d9526d712c6cf38e02708&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 186.0 to 190.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR