(3,3,3-Trifluoropropyl)dimethoxymethylsilane ≥98.0%

Leverantör: TCI
Varning

Synonyms: Dimethoxy(methyl)(3,3,3-trifluoropropyl)silane, 1,1,1-Trifluoro-3-[dimethoxy(methyl)silyl]propane

D5328-5G D5328-25G
TCIAD5328-5GEA 470 SEK
TCIAD5328-5G TCIAD5328-25G
(3,3,3-Trifluoropropyl)dimethoxymethylsilane ≥98.0%
(3,3,3-Trifluoropropyl)dimethoxymethylsilane
Formel: C₆H₁₃F₃O₂Si
Kokpunkt: 85°C
Flampunkt: 58°C
MDL Number: MFCD00053175
CAS nummer: 358-67-8
EINECS: 206-619-4
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=753030e3c7b1448d4dd5f790d65dc6ad8905f987&vid=753030e3c7b1448d4dd5f790d65dc6ad8905f987&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR