2,4-Di-tert-pentylphenol ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 2,4-Di-tert-amylphenol, 2,4-Bis(1,1-dimethylpropyl)phenol, 2,4-Di-tert-pentylphenol
D5320-25G D5320-400G
TCIAD5320-25GEA 245 SEK
TCIAD5320-25G TCIAD5320-400G
2,4-Di-tert-pentylphenol ≥98.0%
2,4-Di-tert-pentylphenol
Formel: C₁₆H₂₆O
MW: 234,38 g/mol
Kokpunkt: 143°C
MDL Number: MFCD00041929
CAS nummer: 120-95-6
EINECS: 204-439-0
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=950607fa02c5eecacaabb14168df426833530fb7&vid=950607fa02c5eecacaabb14168df426833530fb7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Yellow to Green powder to lump to clear liquid
Purity (GC) min. 98.0 %