1,3-Dibromo-5-dodecylbenzene ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
D5274-1G
D5274-5G
TCIAD5274-1GEA
1300
SEK
TCIAD5274-1G
TCIAD5274-5G
1,3-Dibromo-5-dodecylbenzene ≥97.0%
1,3-Dibromo-5-dodecylbenzene
Formel:
C₁₈H₂₈Br₂ MW: 404,23 g/mol Smältpunkt: 53°C Storage Temperature: Kylskåp |
CAS nummer:
231606-27-2 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to lump |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...