2,6-Diiodopyridine ≥97.0%

Leverantör: TCI EUROPE
Varning

D5268-1G D5268-5G
TCIAD5268-1GEA 1400 SEK
TCIAD5268-1G TCIAD5268-5G
2,6-Diiodopyridine ≥97.0%
2,6-Diiodopyridine
Formel: C₅H₃I₂N
MW: 330,89 g/mol
MDL Number: MFCD07366728
CAS nummer: 53710-17-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecb938c14b493b761a2be20e3d4c00f3e1dac623&vid=ecb938c14b493b761a2be20e3d4c00f3e1dac623&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR