3-[[Dimethyl(vinyl)silyl]oxy]-1,1,5,5-tetramethyl-3-phenyl-1,5-divinyltrisiloxane ≥98.0%

Leverantör: TCI
Varning

Synonyms: Tris[dimethyl(vinyl)siloxy]phenylsilane

D5264-5G D5264-25G
TCIAD5264-5GEA 1100 SEK
TCIAD5264-5G TCIAD5264-25G
3-[[Dimethyl(vinyl)silyl]oxy]-1,1,5,5-tetramethyl-3-phenyl-1,5-divinyltrisiloxane ≥98.0%
3-[[Dimethyl(vinyl)silyl]oxy]-1,1,5,5-tetramethyl-3-phenyl-1,5-divinyltrisiloxane
Formel: C₁₈H₃₂O₃Si₄
MW: 408.79 g/mol
MDL Number: MFCD00156495
CAS nummer: 60111-47-9
EINECS: 262-056-4

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR