1,6-Dibromo-3,8-diisopropylpyrene ≥98.0%
Leverantör: TCI EUROPE
D5236-200MG
D5236-1G
TCIAD5236-200MGEA
990
SEK
TCIAD5236-200MG
TCIAD5236-1G
1,6-Dibromo-3,8-diisopropylpyrene ≥98.0%
1,6-Dibromo-3,8-diisopropylpyrene
Formel:
C₂₂H₂₀Br₂ MW: 444,21 g/mol |
MDL Number:
MFCD27979119 CAS nummer: 869340-02-3 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...